Gene Transcripts
- Gene Symbol
- HAR1B
- Entrez Gene ID
- 768097
- Gene Name
- highly accelerated region 1B (non-protein coding)
- Gene Aliases
- HAR1R, LINC00065, NCRNA00065
- Chromosome Location
- Chr. 20: 63095493 - 63102319 on Build GRCh38
- UniGene ID
- Hs.635159
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
HY011775.1 |
- |
1 |
129 |
|
Chr. 20 - 63095871 - 63102309 [-] |
GenBankmRNA |
DB088004.1 |
- |
1 |
99 |
|
Chr. 20 - 63095798 - 63102279 [-] |
GenBankmRNA |
HY023235.1 |
- |
1 |
103 |
|
Chr. 20 - 63095892 - 63102283 [-] |
View Full Product Details