Gene Transcripts
- Gene Symbol
- PEG3-AS1
- Entrez Gene ID
- 100169890
- Gene Name
- PEG3 antisense RNA 1
- Gene Aliases
- APEG3, NCRNA00155, PEG3-AS, PEG3AS
- Chromosome Location
- Chr. 19: 56812480 - 56813793 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023847.2 |
|
1 |
862 |
|
View Full Product Details