Gene Transcripts
- Gene Symbol
- HEXA-AS1
- Entrez Gene ID
- 80072
- Gene Name
- HEXA antisense RNA 1
- Gene Aliases
- C15orf34
- Chromosome Location
- Chr. 15: 72376113 - 72378788 on Build GRCh38
- UniGene ID
- Hs.591113
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027262.1 |
|
1 |
2023 |
|
GenBank mRNA |
BC068211.1 |
- |
1 |
2023 |
|
View Full Product Details