Gene Transcripts
- Gene Symbol
- KTN1-AS1
- Entrez Gene ID
- 100129075
- Gene Name
- KTN1 antisense RNA 1
- Gene Aliases
- C14orf33, MYCLo-3
- Chromosome Location
- Chr. 14: 55576157 - 55580092 on Build GRCh38
- UniGene ID
- Hs.633724
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX362563.2 |
- |
2 |
191 |
|
Chr. 14 - 55576764 - 55580074 [-] |
GenBankmRNA |
BX094019.1 |
- |
2 |
165 |
|
Chr. 14 - 55499681 - 55580062 [-] |
GenBankmRNA |
DA351543.1 |
- |
2 |
207 |
|
Chr. 14 - 55559617 - 55580110 [-] |
GenBankmRNA |
CD675654.1 |
- |
2 |
163 |
|
Chr. 14 - 55577138 - 55580069 [-] |
View Full Product Details