Gene Transcripts

Gene Symbol
POMGNT1
Entrez Gene ID
55624
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2
Chromosome Location
Chr. 1: 46188681 - 46220305 on Build GRCh38
UniGene ID
Hs.525134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243766.1 NP_001230695.1 1 582
GenBank mRNA AK056186.1 - 1 582
CR749700.1 - 1 568


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI919180.1 - 1 221 Chr. 1 - 46196021 - 46219917 [-]
GenBankmRNA HY024680.1 - 2 151 Chr. 1 - 46196833 - 46220980 [-]
GenBankmRNA DA222889.1 - 1 301 Chr. 1 - 46197041 - 46220024 [-]
GenBankmRNA DA222853.1 - 1 301 Chr. 1 - 46197041 - 46220024 [-]
GenBankmRNA DA788644.1 - 1 298 Chr. 1 - 46196987 - 46220021 [-]

View Full Product Details