Gene Transcripts
- Gene Symbol
- POMGNT1
- Entrez Gene ID
- 55624
- Gene Name
- protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
- Gene Aliases
- GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2
- Chromosome Location
- Chr. 1: 46188681 - 46220305 on Build GRCh38
- UniGene ID
- Hs.525134
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA222889.1 |
- |
1 |
23 |
|
Chr. 1 - 46197041 - 46220024 [-] |
GenBankmRNA |
DA222853.1 |
- |
1 |
23 |
|
Chr. 1 - 46197041 - 46220024 [-] |
GenBankmRNA |
DA788644.1 |
- |
1 |
20 |
|
Chr. 1 - 46196987 - 46220021 [-] |
View Full Product Details