Gene Transcripts
- Gene Symbol
- DIO3OS
- Entrez Gene ID
- 64150
- Gene Name
- DIO3 opposite strand/antisense RNA (head to head)
- Gene Aliases
- C14orf134, DIO3-AS1, DIO3-OS, NCRNA00041
- Chromosome Location
- Chr. 14: 101552223 - 101555676 on Build GRCh38
- UniGene ID
- Hs.525597
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000556120.5 |
- |
2 |
2454 |
|
Chr. 14 - 101552223 - 101555676 [-] |
GENCODE |
ENST00000553575.1 |
- |
2 |
966 |
|
Chr. 14 - 101552223 - 101555676 [-] |
GENCODE |
ENST00000557109.5 |
- |
2 |
665 |
|
Chr. 14 - 101552221 - 101560403 [-] |
View Full Product Details