Gene Transcripts

Gene Symbol
C7orf13
Entrez Gene ID
129790
Gene Name
chromosome 7 open reading frame 13
Gene Aliases
MY040
Chromosome Location
Chr. 7: 156638366 - 156640654 on Build GRCh38
UniGene ID
Hs.647014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026865.1 1 1289
GenBank mRNA AF063598.1 - 1 1056
AF439977.1 - 1 1289
AK056371.1 - 1 1205
BC031037.1 - 1 1174

View Full Product Details