Gene Transcripts
- Gene Symbol
- RAET1K
- Entrez Gene ID
- 646024
- Gene Name
- retinoic acid early transcript 1K pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 6: 149998019 - 150005144 on Build GRCh38
- UniGene ID
- Hs.560537
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024045.1 |
|
2 |
453 |
|
GenBank mRNA |
AF425244.1 |
- |
2 |
453 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AI017460.1 |
- |
2 |
435 |
|
Chr. 6 - 150001259 - 150005126 [-] |
View Full Product Details