Gene Transcripts
- Gene Symbol
- NBPF22P
- Entrez Gene ID
- 285622
- Gene Name
- neuroblastoma breakpoint family member 22, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 86282444 - 86297547 on Build GRCh38
- UniGene ID
- Hs.449272
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003719.2 |
|
8 |
1871 |
|
GenBank mRNA |
BC050328.1 |
- |
8 |
1827 |
|
- Gene Symbol
- NBPF4
- Entrez Gene ID
- 148545
- Gene Name
- neuroblastoma breakpoint family member 4
- Gene Aliases
- Chromosome Location
- Chr. 1: 108222464 - 108244081 on Build GRCh38
- UniGene ID
- Hs.511849
- Species
- Homo sapiens
- Gene Symbol
- NBPF6
- Entrez Gene ID
- 653149
- Gene Name
- neuroblastoma breakpoint family member 6
- Gene Aliases
- Chromosome Location
- Chr. 1: 108421496 - 108471903 on Build GRCh38
- UniGene ID
- Hs.712226
- Species
- Homo sapiens
View Full Product Details