Gene Transcripts
- Gene Symbol
- TOP1P2
- Entrez Gene ID
- 7152
- Gene Name
- topoisomerase (DNA) I pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 24764501 - 24766021 on Build GRCh38
- UniGene ID
- Hs.381281
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001283.1 |
|
1 |
217 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
M55630.3 |
- |
1 |
218 |
|
Chr. 22 - 24764501 - 24766156 [+] |
View Full Product Details