Gene Transcripts
- Gene Symbol
- CXADRP2
- Entrez Gene ID
- 646243
- Gene Name
- coxsackie virus and adenovirus receptor pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 15: 21371493 - 21373968 on Build GRCh38
- UniGene ID
- Hs.720910
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024387.1 |
|
1 |
1441 |
|
View Full Product Details