Gene Transcripts

Gene Symbol
COL6A4P2
Entrez Gene ID
646300
Gene Name
collagen type VI alpha 4 pseudogene 2
Gene Aliases
COL6A4
Chromosome Location
Chr. 3: 130212820 - 130273806 on Build GRCh38
UniGene ID
Hs.170475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027898.1 3, 4 152
GenBank mRNA CN285077.1 - 1, 2 57
EG328528.1 - 1, 2 64

View Full Product Details