Gene Transcripts
- Gene Symbol
- POGLUT1
- Entrez Gene ID
- 56983
- Gene Name
- protein O-glucosyltransferase 1
- Gene Aliases
- C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
- Chromosome Location
- Chr. 3: 119468938 - 119494708 on Build GRCh38
- UniGene ID
- Hs.231750
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024265.1 |
|
4, 5 |
548 |
|
GenBank mRNA |
AK295984.1 |
- |
4, 5 |
523 |
|
CD517010.1 |
- |
4, 5 |
464 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DC337928.1 |
- |
4, 5 |
523 |
|
Chr. 3 - 119468963 - 119479967 [+] |
View Full Product Details