Gene Transcripts

Gene Symbol
POGLUT1
Entrez Gene ID
56983
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
C3orf9, CLP46, KDELCL1, KTELC1, MDS010, MDSRP, Rumi, hCLP46
Chromosome Location
Chr. 3: 119468938 - 119494708 on Build GRCh38
UniGene ID
Hs.231750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024265.1 5 585
XM_017006878.1 XP_016862367.1 5 557
XM_017006879.1 XP_016862368.1 6 703
GenBank mRNA AK295984.1 - 5 560
AK301002.1 - 6 701
CD517010.1 - 5 501


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DC337928.1 - 5 560 Chr. 3 - 119468963 - 119479967 [+]

View Full Product Details