Gene Transcripts
- Gene Symbol
- CTAGE10P
- Entrez Gene ID
- 220429
- Gene Name
- CTAGE family member 10, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 13: 49890409 - 49893380 on Build GRCh38
- UniGene ID
- Hs.448219
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003268.3 |
|
1 |
2694 |
|
GenBank mRNA |
BC030655.1 |
- |
1 |
2593 |
|
View Full Product Details