Gene Transcripts
- Gene Symbol
- MIMT1
- Entrez Gene ID
- 100073347
- Gene Name
- MER1 repeat containing imprinted transcript 1 (non-protein coding)
- Gene Aliases
- LINC00067, MIM1, NCRNA00067
- Chromosome Location
- Chr. 19: 56840902 - 56848554 on Build GRCh38
- UniGene ID
- Hs.467337
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024059.1 |
|
2 |
895 |
|
GenBank mRNA |
EF110915.1 |
- |
2 |
895 |
|
View Full Product Details