Gene Transcripts

Gene Symbol
DLX6-AS1
Entrez Gene ID
285987
Gene Name
DLX6 antisense RNA 1
Gene Aliases
DLX6-AS, DLX6AS, Evf-2, NCRNA00212
Chromosome Location
Chr. 7: 96968515 - 97014065 on Build GRCh38
UniGene ID
Hs.34969
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015448.1 2 448
GenBank mRNA AK095619.1 - 2 448
AK308769.1 - 2 365
AK309768.1 - 2 532


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000458352.2 - 2 532 Chr. 7 - 96978468 - 97012049 [-]
GENCODE ENST00000430027.3 - 2 448 Chr. 7 - 96955141 - 97014065 [-]

View Full Product Details