Gene Transcripts

Gene Symbol
TCAM1P
Entrez Gene ID
146771
Gene Name
testicular cell adhesion molecule 1, pseudogene
Gene Aliases
TCAM1
Chromosome Location
Chr. 17: 63857016 - 63864379 on Build GRCh38
UniGene ID
Hs.629177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002947.2 3 972
GenBank mRNA AB026156.1 - 3 972
LC005096.1 - 5 1063
LC005097.1 - 5 832
LC005098.1 - 4 800
LC005099.1 - 5 917
LC005100.1 - 5 848
LC005101.1 - 5 814

View Full Product Details