Gene Transcripts
- Gene Symbol
- PGCP1
- Entrez Gene ID
- 441897
- Gene Name
- progastricsin pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 111384519 - 111389851 on Build GRCh38
- UniGene ID
- Hs.568387
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029429.1 |
|
1 |
20 |
|
View Full Product Details