Gene Transcripts

Gene Symbol
NIPSNAP3B
Entrez Gene ID
55335
Gene Name
nipsnap homolog 3B
Gene Aliases
FP944, NIPSNAP3, SNAP1
Chromosome Location
Chr. 9: 104763741 - 104777764 on Build GRCh38
UniGene ID
Hs.429294
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018376.3 NP_060846.2 6 5529
NR_130759.1 5 5318
NR_130760.1 6 1428
XM_011518839.1 XP_011517141.1 5 5379
XM_017014889.1 XP_016870378.1 6 1350
GenBank mRNA AF370389.1 - 1 1817
AK091667.1 - 1 1822
BX107655.1 - 2 327

View Full Product Details