Gene Transcripts
- Gene Symbol
- CYCSP52
- Entrez Gene ID
- 360155
- Gene Name
- cytochrome c, somatic pseudogene 52
- Gene Aliases
- HC6, HCP2
- Chromosome Location
- Chr. 1: 157128362 - 157128671 on Build GRCh38
- UniGene ID
- Hs.491808
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001560.1 |
|
1 |
31 |
|
GenBank mRNA |
R91214.1 |
- |
1 |
113 |
|
View Full Product Details