Gene Transcripts

Gene Symbol
PRAC2
Entrez Gene ID
360205
Gene Name
prostate cancer susceptibility candidate 2
Gene Aliases
C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253
Chromosome Location
Chr. 17: 48720802 - 48724758 on Build GRCh38
UniGene ID
Hs.236557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282275.1 NP_001269204.1 2 198
NM_001282276.1 NP_001269205.1 2 132
XM_006721865.3 XP_006721928.1 2 814
XM_011524747.2 XP_011523049.1 2 124
XM_011524748.1 XP_011523050.1 2 69
GenBank mRNA AF532777.1 - 2 186
BX106051.1 - 2 132


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB048981.1 - 2 162 Chr. 17 - 48723209 - 48724729 [+]

View Full Product Details