Gene Transcripts
- Gene Symbol
- RMST
- Entrez Gene ID
- 196475
- Gene Name
- rhabdomyosarcoma 2 associated transcript (non-protein coding)
- Gene Aliases
- LINC00054, NCRMS, NCRNA00054
- Chromosome Location
- Chr. 12: 97465021 - 97533766 on Build GRCh38
- UniGene ID
- Hs.652568
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA746072.1 |
- |
2 |
148 |
|
Chr. 12 - 97493950 - 97530844 [+] |
GenBankmRNA |
DA746073.1 |
- |
1 |
150 |
|
Chr. 12 - 97494742 - 97530844 [+] |
View Full Product Details