Gene Transcripts
- Gene Symbol
- RPPH1
- Entrez Gene ID
- 85495
- Gene Name
- ribonuclease P RNA component H1
- Gene Aliases
- H1RNA, RPPH1-1
- Chromosome Location
- Chr. 14: 20343071 - 20343411 on Build GRCh38
- UniGene ID
- Hs.613102
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002312.1 |
|
1 |
81 |
|
GenBank mRNA |
DW429001.1 |
- |
1 |
97 |
|
X15624.1 |
- |
1 |
80 |
|
View Full Product Details