Gene Transcripts

Gene Symbol
SCARNA10
Entrez Gene ID
692148
Gene Name
small Cajal body-specific RNA 10
Gene Aliases
U85
Chromosome Location
Chr. 12: 6510222 - 6510551 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_004387.1 1 160


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ069333.1 - 64 Chr. - - [-]

View Full Product Details