Gene Transcripts
- Gene Symbol
- SCARNA10
- Entrez Gene ID
- 692148
- Gene Name
- small Cajal body-specific RNA 10
- Gene Aliases
- U85
- Chromosome Location
- Chr. 12: 6510222 - 6510551 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004387.1 |
|
1 |
27 |
|
View Full Product Details