Gene Transcripts
- Gene Symbol
- Six3os1
- Entrez Gene ID
- 100043902
- Gene Name
- SIX homeobox 3, opposite strand 1
- Gene Aliases
- D17Mgi26, E130112H22Rik, Rncr1, Six3os
- Chromosome Location
- Chr. 17: 85601932 - 85618396 on Build GRCm38
- UniGene ID
- Mm.150838
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK044330.1 |
- |
LIT3371 RNCR1 |
2 |
847 |
|
View Full Product Details