Gene Transcripts

Gene Symbol
Mir155hg
Entrez Gene ID
100653389
Gene Name
uncharacterized LOC100653389
Gene Aliases
Bic
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.261808
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY096003.1 - LIT1587
bic
2 721

View Full Product Details