Gene Transcripts

Gene Symbol
MIR18A
Entrez Gene ID
406953
Gene Name
microRNA 18a
Gene Aliases
MIR18, MIRN18, MIRN18A, hsa-mir-18, hsa-mir-18a, miR-18, miRNA18A, mir-18a
Chromosome Location
Chr. 13: 91350751 - 91350821 on Build GRCh38
UniGene ID
Hs.24115
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB176708.1 - LIT3364
oncomir-1
2 5023

View Full Product Details