Gene Transcripts

Gene Symbol
SNRPN
Entrez Gene ID
6638
Gene Name
small nuclear ribonucleoprotein polypeptide N
Gene Aliases
HCERN3, PWCR, RT-LI, SM-D, SMN, SNRNP-N, SNURF-SNRPN, sm-N
Chromosome Location
Chr. 15: 24823637 - 24978723 on Build GRCh38
UniGene ID
Hs.555970
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA U81001.1 - LIT2197
IC-SNURF-SNRPN 3 prime noncoding transcripts
1 566

View Full Product Details