Gene Transcripts
- Gene Symbol
- PBDC1
- Entrez Gene ID
- 51260
- Gene Name
- polysaccharide biosynthesis domain containing 1
- Gene Aliases
- CXorf26
- Chromosome Location
- Chr. X: 76172929 - 76178310 on Build GRCh38
- UniGene ID
- Hs.370100
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927616.1 |
- |
LIT3604 AY927616 |
|
11 |
|
View Full Product Details