Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
22931
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
RAB18LI1, WARBM3
Chromosome Location
Chr. 10: 27504174 - 27542237 on Build GRCh38
UniGene ID
Hs.406799
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256410.1 NP_001243339.1 8 4248
NM_001256411.1 NP_001243340.1 6 4094
NM_001256412.1 NP_001243341.1 5 3969
NM_001256415.1 NP_001243344.1 6 4089
NM_021252.4 NP_067075.1 7 4161
NR_046172.1 6 4099


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR610854.1 - 1 869 Chr. 10 - 27540527 - 27542214 [+]

View Full Product Details