Gene Transcripts
- Gene Symbol
- IFT52
- Entrez Gene ID
- 51098
- Gene Name
- intraflagellar transport 52
- Gene Aliases
- C20orf9, CGI-53, NGD2, NGD5
- Chromosome Location
- Chr. 20: 43590613 - 43647296 on Build GRCh38
- UniGene ID
- Hs.444332
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097603.1 |
- |
2 |
1386 |
|
View Full Product Details