Gene Transcripts
- Gene Symbol
- SLC26A6
- Entrez Gene ID
- 65010
- Gene Name
- solute carrier family 26 member 6
- Gene Aliases
- Chromosome Location
- Chr. 3: 48625723 - 48635493 on Build GRCh38
- UniGene ID
- Hs.631925
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF161369.1 |
- |
1 |
726 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF150415.1 |
- |
1 |
726 |
|
Chr. 19 - 15273835 - 15274702 [-] |
View Full Product Details