Gene Transcripts

Gene Symbol
PC
Entrez Gene ID
5091
Gene Name
pyruvate carboxylase
Gene Aliases
PCB
Chromosome Location
Chr. 11: 66848522 - 66958418 on Build GRCh38
UniGene ID
Hs.89890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017017872.1 XP_016873361.1 1 429
GenBank mRNA U30890.1 - 1 46

View Full Product Details