Gene Transcripts

Gene Symbol
NAG20
Entrez Gene ID
100129703
Gene Name
NAG20
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.664650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF210649.1 - 1 610

View Full Product Details