Gene Transcripts

Gene Symbol
FCF1
Entrez Gene ID
51077
Gene Name
FCF1 rRNA-processing protein
Gene Aliases
Bka, C14orf111, CGI-35, UTP24
Chromosome Location
Chr. 14: 74713118 - 74738620 on Build GRCh38
UniGene ID
Hs.579828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005267730.3 XP_005267787.1 8 4215
GenBank mRNA AK057060.1 - 1 1608

View Full Product Details