Gene Transcripts
- Gene Symbol
- TIMM44
- Entrez Gene ID
- 10469
- Gene Name
- translocase of inner mitochondrial membrane 44
- Gene Aliases
- TIM44
- Chromosome Location
- Chr. 19: 7926718 - 7943823 on Build GRCh38
- UniGene ID
- Hs.465784
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK309415.1 |
- |
5 |
379 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BC022423.1 |
- |
1 |
678 |
|
Chr. 19 - 7935168 - 7935880 [-] |
View Full Product Details