Gene Transcripts

Gene Symbol
ZBTB20
Entrez Gene ID
26137
Gene Name
zinc finger and BTB domain containing 20
Gene Aliases
DPZF, HOF, ODA-8S, PRIMS, ZNF288
Chromosome Location
Chr. 3: 114314500 - 115147280 on Build GRCh38
UniGene ID
Hs.202577
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164342.2 NP_001157814.1 5 4268
NM_001164343.2 NP_001157815.1 12 4730
NM_001164344.2 NP_001157816.1 8 4694
NM_001164345.2 NP_001157817.1 7 4648
NM_001164346.2 NP_001157818.1 6 4287
NM_001164347.2 NP_001157819.1 6 4309
NM_015642.5 NP_056457.3 10 4492
NR_121662.1 7 2750
GenBank mRNA AK022228.1 - 1 109

View Full Product Details