Gene Transcripts
- Gene Symbol
- PKI55
- Entrez Gene ID
- 150967
- Gene Name
- DKFZp434H1419
- Gene Aliases
- Chromosome Location
- Chr. 2: 216216889 - 216220192 on Build GRCh38
- UniGene ID
- Hs.56876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037701.1 |
|
1 |
3045 |
|
GenBank mRNA |
AK125369.1 |
- |
1 |
2889 |
|
AL137534.1 |
- |
1 |
2353 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB563055.1 |
- |
1 |
165 |
|
Chr. 2 - 216219770 - 216220192 [+] |
GenBankmRNA |
DB573280.1 |
- |
1 |
213 |
|
Chr. 2 - 216219721 - 216220192 [+] |
GenBankmRNA |
DB575568.1 |
- |
1 |
226 |
|
Chr. 2 - 216219708 - 216220192 [+] |
View Full Product Details