Gene Transcripts

Gene Symbol
PKI55
Entrez Gene ID
150967
Gene Name
DKFZp434H1419
Gene Aliases
Chromosome Location
Chr. 2: 216216889 - 216220192 on Build GRCh38
UniGene ID
Hs.56876
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037701.1 1 1723
GenBank mRNA AK125369.1 - 1 1567
AL137534.1 - 1 1027


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DN991765.1 - 1 339 Chr. 2 - 216218281 - 216218922 [+]
GenBankmRNA BX490494.1 - 1 413 Chr. 2 - 216218199 - 216218839 [+]
GenBankmRNA DA871533.1 - 1 354 Chr. 2 - 216218258 - 216218797 [+]
GenBankmRNA DA166487.1 - 1 317 Chr. 2 - 216218295 - 216218860 [+]
GenBankmRNA DA715983.1 - 1 313 Chr. 2 - 216218299 - 216218852 [+]

View Full Product Details