Gene Transcripts

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
Chromosome Location
Chr. 6: 16299112 - 16761490 on Build GRCh38
UniGene ID
Hs.434961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311649.1 - 2 792
BC011026.1 - 2 825
BC014548.1 - 3 894
BC039236.1 - 3 817
BC047894.1 - 3 923
BC063120.1 - 3 918

View Full Product Details