Gene Transcripts

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
Chromosome Location
Chr. 6: 16299112 - 16761490 on Build GRCh38
UniGene ID
Hs.434961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311649.1 - 2 451
BC011026.1 - 2 484
BC014548.1 - 3 553
BC039236.1 - 3 476
BC047894.1 - 3 582
BC063120.1 - 3 577


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BU602093.1 - 3 576 Chr. 6 - 16739475 - 16761469 [-]
GenBankmRNA DA975674.1 - 2 416 Chr. 6 - 16739485 - 16753347 [-]
GenBankmRNA BG253572.1 - 3 581 Chr. 6 - 16739511 - 16761475 [-]
GenBankmRNA BM919920.1 - 3 578 Chr. 6 - 16739237 - 16761439 [-]
GenBankmRNA DA415596.1 - 4 617 Chr. 6 - 16739602 - 16761459 [-]
GenBankmRNA BG497560.1 - 3 474 Chr. 6 - 16739282 - 16761371 [-]

View Full Product Details