Gene Transcripts

Gene Symbol
SLIT3
Entrez Gene ID
6586
Gene Name
slit guidance ligand 3
Gene Aliases
MEGF5, SLIL2, SLIT1, Slit-3, slit2
Chromosome Location
Chr. 5: 168661733 - 169301129 on Build GRCh38
UniGene ID
Hs.552087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271946.1 NP_001258875.1 36 8803
NM_003062.3 NP_003053.1 36 8782
XM_017009779.1 XP_016865268.1 36 8185
GenBank mRNA BC062365.1 - 1 516


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA ES313324.1 - 1 127 Chr. 5 - 168662247 - 168662790 [-]
GenBankmRNA CN281115.1 - 1 318 Chr. 5 - 168662205 - 168662973 [-]
GenBankmRNA CN281116.1 - 1 277 Chr. 5 - 168662204 - 168662933 [-]
GenBankmRNA BQ719154.1 - 1 155 Chr. 5 - 168661956 - 168662818 [-]
GenBankmRNA BQ070258.1 - 1 512 Chr. 5 - 168662279 - 168663176 [-]

View Full Product Details