Gene Transcripts
- Gene Symbol
- SELK
- Entrez Gene ID
- 58515
- Gene Name
- selenoprotein K
- Gene Aliases
- HSPC030, HSPC297, SelK
- Chromosome Location
- Chr. 3: 53885199 - 53891962 on Build GRCh38
- UniGene ID
- Hs.58471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123406.1 |
- |
1 |
2130 |
|
View Full Product Details