Gene Transcripts
- Gene Symbol
- HELLS
- Entrez Gene ID
- 3070
- Gene Name
- helicase, lymphoid-specific
- Gene Aliases
- ICF4, LSH, Nbla10143, PASG, SMARCA6
- Chromosome Location
- Chr. 10: 94545767 - 94613905 on Build GRCh38
- UniGene ID
- Hs.463677
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK021443.1 |
- |
1 |
1346 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BP422666.1 |
- |
1 |
389 |
|
Chr. 10 - 94613243 - 94613775 [+] |
View Full Product Details