Gene Transcripts

Gene Symbol
L3MBTL4
Entrez Gene ID
91133
Gene Name
l(3)mbt-like 4 (Drosophila)
Gene Aliases
HsT1031
Chromosome Location
Chr. 18: 5954703 - 6414911 on Build GRCh38
UniGene ID
Hs.128279
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011525762.1 XP_011524064.1 17 3032
GenBank mRNA AK098363.1 - 1 547

View Full Product Details