Gene Transcripts
- Gene Symbol
- LRRC8B
- Entrez Gene ID
- 23507
- Gene Name
- leucine rich repeat containing 8 family member B
- Gene Aliases
- TA-LRRP, TALRRP
- Chromosome Location
- Chr. 1: 89522627 - 89597861 on Build GRCh38
- UniGene ID
- Hs.482017
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC053565.1 |
- |
1 |
210 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ717287.1 |
- |
1 |
209 |
|
Chr. 1 - 89584150 - 89584917 [-] |
View Full Product Details