Gene Transcripts
- Gene Symbol
- IQCA1
- Entrez Gene ID
- 79781
- Gene Name
- IQ motif containing with AAA domain 1
- Gene Aliases
- 4930465P12Rik, DRC11, IQCA
- Chromosome Location
- Chr. 2: 236324147 - 236507558 on Build GRCh38
- UniGene ID
- Hs.591594
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC018494.2 |
- |
1 |
449 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG249408.1 |
- |
|
461 |
|
Chr. - - [-] |
View Full Product Details